APO455838576 Apollo Scientific Ltd

4,5-Dichloroperfluoro(2,2-dimethyl-1,3-dioxolane)

  • MDL number MFCD01937893
  • Cas Number : 60644-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2-Bis(trifluoromethyl)-4,5-dichloro-4,5-difluoro-1,3-dioxolane
Density 1.587
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 88-90
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47480

Safety Information

signalword Warning
pcodes P210;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47480
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart