APO455838579 Apollo Scientific Ltd

1-[6-Fluoro-2-(trifluoromethyl)quinolin-4-yl]homopiperazine 250mg pack

  • MDL number MFCD03094525
  • Cas Number : 541539-67-7
  • Molecular Weight : 313.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-6-fluoro-2-trifluoromethylquinolin-4-ylhomopiperazine

Safety Information

signalword Warning
pcodes P271;P261;P280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart