APO455838588 Apollo Scientific Ltd

2-(Aminomethyl)-6-fluoropyridine dihydrochloride

  • MDL number MFCD16872044
  • Cas Number : 1257535-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (6-Fluoropyridin-2-yl)methylamine dihydrochloride, 6-Fluoropicolinylamine dihydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47684

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47684
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart