APO455838589 Apollo Scientific Ltd

4-Bromo-2-fluorophenylacetonitrile

  • MDL number MFCD07368653
  • Cas Number : 114897-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-53
Synonym 4-Bromo-2-fluorobenzyl cyanide
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-fluorophenylacetonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-fluorophenylacetonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart