APO455838590 Apollo Scientific Ltd

4-(Aminomethyl)-3-fluoropyridine dihydrochloride

  • MDL number MFCD12922842
  • Cas Number : 1257535-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47682

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=47682
pictograms GHS05;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart