APO455838592 Apollo Scientific Ltd

3-Fluoroazetidine hydrochloride

  • MDL number MFCD05663715
  • Cas Number : 617718-46-4
  • Molecular Weight : 111.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Fluoroazetane hydrochloride
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoroazetidine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoroazetidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart