APO455838605 Apollo Scientific Ltd

2-Chloro-5-(trifluoromethyl)isonicotinic acid

  • MDL number MFCD01862019
  • Cas Number : 505084-58-2
  • Molecular Weight : 225.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186(dec.)
Synonym 2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-trifluoromethylisonicotinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-trifluoromethylisonicotinic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart