APO455838608 Apollo Scientific Ltd

2,3,5,6-Tetrafluorobenzoic acid

  • MDL number MFCD00002408
  • Cas Number : 652-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-154
Synonym 4H-Tetrafluorobenzoic acid
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2356-tetrafluorobenzoic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2356-tetrafluorobenzoic-acid
pictograms GHS05
hstatements H315;H318
Availability details for
Quantity
  • Availability
  • Add to Cart