APO455838614 Apollo Scientific Ltd

1-Acetyl-3,5-bis(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD00153613
  • Cas Number : 244187-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=9
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart