APO455838616 Apollo Scientific Ltd

5,7-Bis(trifluoromethyl)-3-cyanopyrazolo[1,5-a]pyrimidine

  • MDL number MFCD00084901
  • Cas Number : 175276-40-1
  • Molecular Weight : 280.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-77
Purity 97
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/57-bistrifluoromethyl-3-cyanopyrazolo15-apyrimidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/57-bistrifluoromethyl-3-cyanopyrazolo15-apyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart