APO455838619 Apollo Scientific Ltd

5-[3,5-Bis(trifluoromethyl)phenyl]-2,4-dihydro-3H-1,2,4-triazole-3-thione

  • MDL number MFCD00177275
  • Cas Number : 175276-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280
Synonym 3-[3,5-Bis(trifluoromethyl)phenyl]-4,5-dihydro-5-thioxo-1H-1,2,4-triazole
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=392

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=392
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart