APO455838622 Apollo Scientific Ltd

4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluorononan-1-ol

  • MDL number MFCD00077578
  • Cas Number : 80806-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -3
Synonym 3-(Perfluorohex-1-yl)propan-1-ol, 1H,1H,2H,2H,3H,3H-Perfluorononan-1-ol
Density 1.629
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 70-80/10mm
Product Link https://store.apolloscientific.co.uk/product/4455667788999-tridecafluorononan-1-ol

Safety Information

signalword Warning
pcodes P210;P241;P280
MSDS https://store.apolloscientific.co.uk/product/4455667788999-tridecafluorononan-1-ol
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart