APO455838624 Apollo Scientific Ltd

3,3-Difluoro-3-(heptafluoropropyl)-1,2-propenoxide

  • MDL number MFCD00042341
  • Cas Number : 89807-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Perfluorobut-1-yl)oxirane, 1-(Oxiran-2-yl)perfluorobutane
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 52/41mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4173

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4173
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart