APO455838627 Apollo Scientific Ltd

2,4-Bis(trifluoromethyl)phenylacetonitrile

  • MDL number MFCD00042490
  • Cas Number : 201789-28-8
  • Molecular Weight : 253.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-bistrifluoromethylphenylacetonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-bistrifluoromethylphenylacetonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart