APO455838630 Apollo Scientific Ltd

2-Fluorobenzoylacetonitrile

  • MDL number MFCD02260800
  • Cas Number : 31915-26-1
  • Molecular Weight : 163.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-62
Synonym 3-(2-Fluorophenyl)-3-oxopropanenitrile
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluorobenzoylacetonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluorobenzoylacetonitrile
pictograms GHS07
hstatements H302+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart