APO455838633 Apollo Scientific Ltd

2-(Trifluoromethoxy)benzoylacetonitrile

  • MDL number MFCD08458117
  • Cas Number : 914636-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Oxo-3-[2-(trifluoromethoxy)phenyl]propanenitrile, 3-Oxo-3-[2-(trifluoromethoxy)phenyl]propionitrile
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethoxybenzoylacetonitrile-98

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethoxybenzoylacetonitrile-98
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart