APO455838635 Apollo Scientific Ltd

2-(Trifluoromethoxy)phenylacetic acid

  • MDL number MFCD00236325
  • Cas Number : 220239-67-8
  • Molecular Weight : 220.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-56
Synonym 2-(Carboxymethyl)-alpha,alpha,alpha-trifluoroanisole
Purity 98
Flash Point 103
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethoxyphenylacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethoxyphenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart