APO455838637 Apollo Scientific Ltd

5-Chloro-2,4-difluorobromobenzene

  • MDL number MFCD04112557
  • Cas Number : 914636-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-5-chloro-2,4-difluorobenzene
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-chloro-24-difluorobromobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-24-difluorobromobenzene
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart