APO455838640 Apollo Scientific Ltd

3-Fluoro-4-hydroxybenzaldehyde

  • MDL number MFCD00016626
  • Cas Number : 405-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-125
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1818

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=1818
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart