APO455838641 Apollo Scientific Ltd

1,5-Dibromo-2,4-difluorobenzene

  • MDL number MFCD08458122
  • Cas Number : 28342-75-8
  • Molecular Weight : 271.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29-31
Purity 98
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/15-dibromo-24-difluorobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/15-dibromo-24-difluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart