APO455838644 Apollo Scientific Ltd

3-Fluoro-5-iodobenzaldehyde

  • MDL number MFCD07779030
  • Cas Number : 914636-93-4
  • Molecular Weight : 250.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-5-iodobenzaldehyde
Inchi Code 1S/C7H4FIO/c8-6-1-5(4-10)2-7(9)3-6/h1-4H
Inchi Key XULKBHBAGSJSFP-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-iodobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-iodobenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart