APO455838646 Apollo Scientific Ltd

2,3,4,6-Tetrafluorobenzoic acid

  • MDL number MFCD00039523
  • Cas Number : 32890-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-98
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4777

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=4777
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart