APO455838652 Apollo Scientific Ltd

4-Fluoro-3-methoxyaniline

  • MDL number MFCD00665789
  • Cas Number : 64465-53-8
  • Molecular Weight : 141.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-68
Synonym 5-Amino-2-fluoroanisole, 4-Fluoro-m-anisidine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-methoxyaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-methoxyaniline
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart