APO455838655 Apollo Scientific Ltd

1-[3-(Trifluoromethyl)pyridin-2-yl]piperazine

  • MDL number MFCD00114705
  • Cas Number : 87394-63-6
  • Molecular Weight : 231.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-47
Synonym 2-(Piperazin-1-yl)-3-(trifluoromethyl)pyridine
Purity 99
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 106/1.5mm
Product Link https://store.apolloscientific.co.uk/product/1-3-trifluoromethylpyridin-2-ylpiperazine

Safety Information

signalword Danger
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/1-3-trifluoromethylpyridin-2-ylpiperazine
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart