APO455838656 Apollo Scientific Ltd

3-Fluoro-4-(trifluoromethyl)benzaldehyde

  • MDL number MFCD00236275
  • Cas Number : 204339-72-0
  • Molecular Weight : 192.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha,alpha,alpha,3-Tetrafluoro-p-tolualdehyde, 2-Fluoro-4-formylbenzotrifluoride
Purity 97
Flash Point 73
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-trifluoromethylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-trifluoromethylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart