APO455838657 Apollo Scientific Ltd

3-Bromo-2-(trifluoromethyl)pyridine

  • MDL number MFCD08458124
  • Cas Number : 590371-58-7
  • Molecular Weight : 226

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-43
Synonym 3-Bromo-alpha,alpha,alpha-trifluoro-2-picoline
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 97-99/26mm
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart