APO455838663 Apollo Scientific Ltd

2-Chloro-3-methyl-1,1,2-trifluoro-3-vinylcyclobutane

  • MDL number MFCD00039438
  • Cas Number : 4265-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-1-ethenyl-1-methyl-2,3,3-trifluorocyclobutane
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5078

Safety Information

signalword Warning
pcodes P210;P241;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5078
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart