APO455838666 Apollo Scientific Ltd

4-Fluoro-3-nitrophenyl methyl sulphone

  • MDL number MFCD00025066
  • Cas Number : 453-72-5
  • Molecular Weight : 219.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-157
Synonym 2-Fluoro-5-(methylsulphonyl)nitrobenzene
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-nitrophenyl-methyl-sulphone

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-nitrophenyl-methyl-sulphone
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart