APO455838667 Apollo Scientific Ltd

2-Bromo-4-(trifluoromethyl)benzeneboronic acid

  • MDL number MFCD08532500
  • Cas Number : 959997-88-7
  • Molecular Weight : 268.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Borono-3-bromobenzotrifluoride
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-trifluoromethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-trifluoromethylbenzeneboronic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart