APO455838668 Apollo Scientific Ltd

4-Methyl-3-nitrobenzotrifluoride

  • MDL number MFCD00272402
  • Cas Number : 65754-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Methyl-2-nitro-4-(trifluoromethyl)benzene, 2-Methyl-5-(trifluoromethyl)nitrobenzene, 3-Nitro-alpha,alpha,alpha-trifluoro-p-xylene
Purity 98
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 76-78/8mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11021

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=11021
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart