APO455838673 Apollo Scientific Ltd

4-Bromo-2,3-difluorobenzaldehyde

  • MDL number MFCD09258953
  • Cas Number : 644985-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-65
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11249

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=11249
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart