APO455838674 Apollo Scientific Ltd

4-Bromo-2,3-difluorobenzoic acid

  • MDL number MFCD07368121
  • Cas Number : 194804-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-196
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11250

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=11250
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart