APO455838682 Apollo Scientific Ltd

2-Amino-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD00519334
  • Cas Number : 402-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Synonym 4-(Trifluoromethyl)anthranilic acid, 4-Carboxy-alpha,alpha,alpha-trifluoro-m-toluidine
Purity 98
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-trifluoromethylbenzoic-acid-98

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-trifluoromethylbenzoic-acid-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart