APO455838683 Apollo Scientific Ltd

3-Amino-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD07784307
  • Cas Number : 125483-00-3
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198.6-205
Synonym 5-Carboxy-2-(trifluoromethyl)aniline, 2-Amino-4-carboxybenzotrifluoride
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart