APO455838691 Apollo Scientific Ltd

3-Fluoro-4-(trifluoromethyl)phenylacetonitrile

  • MDL number MFCD00236305
  • Cas Number : 247113-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Fluoro-4-(trifluoromethyl)benzyl cyanide, 4-(Cyanomethyl)-2-fluorobenzotrifluoride
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2232

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=2232
pictograms GHS06;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart