APO455838692 Apollo Scientific Ltd

4-Fluoro-3-iodobenzoic acid

  • MDL number MFCD03788556
  • Cas Number : 403-18-9
  • Molecular Weight : 266.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 173-175
Iupac Name 4-fluoro-3-iodobenzoic acid
Inchi Code 1S/C7H4FIO2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3H,(H,10,11)
Inchi Key IKSOHJRYPQRTLQ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 173-175
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-iodobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-iodobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart