APO455838697 Apollo Scientific Ltd

N-Cyclopropyl-N-(piperidin-4-yl)-3-(trifluoromethyl)benzenesulphonamide

  • MDL number MFCD01568000
  • Cas Number : 387350-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >195(dec.)
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2906

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=2906
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart