APO455838700 Apollo Scientific Ltd

2-Fluoro-6-hydroxybenzaldehyde

  • MDL number MFCD01090996
  • Cas Number : 38226-10-7
  • Molecular Weight : 140.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-39
Synonym 6-Fluorosalicylaldehyde, 3-Fluoro-2-formylphenol
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-hydroxybenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-hydroxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart