APO455838702 Apollo Scientific Ltd

3-Cyano-5-nitrobenzotrifluoride

  • MDL number MFCD03412207
  • Cas Number : 20566-80-7
  • Molecular Weight : 216.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-nitro-5-(trifluoromethyl)benzonitrile
Inchi Code 1S/C8H3F3N2O2/c9-8(10,11)6-1-5(4-12)2-7(3-6)13(14)15/h1-3H
Inchi Key FSQINZQIPFZWKX-UHFFFAOYSA-N

Description

Synonym 3-Nitro-5-(trifluoromethyl)benzonitrile
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-cyano-5-nitrobenzotrifluoride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-cyano-5-nitrobenzotrifluoride
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart