APO455838703 Apollo Scientific Ltd

Decafluorodiphenyl sulphide

  • MDL number MFCD00000293
  • Cas Number : 1043-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-88
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2908

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=2908
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart