APO455838709 Apollo Scientific Ltd

1-Fluoro-4-(tributylstannyl)benzene

  • MDL number MFCD01313817
  • Cas Number : 17151-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 132-134/1mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2112

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=2112
pictograms GHS06;GHS08;GHS09
Availability details for
Quantity
  • Availability
  • Add to Cart