APO455838713 Apollo Scientific Ltd

2-Fluoro-6-methylbenzyl bromide

  • MDL number MFCD06660281
  • Cas Number : 886502-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha'-Bromo-3-fluoro-o-xylene, 2-(Bromomethyl)-1-fluoro-3-methylbenzene
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=33455

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=33455
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart