APO455838720 Apollo Scientific Ltd

4-Bromo-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD08459288
  • Cas Number : 161622-14-6
  • Molecular Weight : 269.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-5-carboxybenzotrifluoride, 4-Bromo-alpha,alpha,alpha-trifluoro-m-toluic acid
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart