APO455838724 Apollo Scientific Ltd

4-(Trifluoromethyl)phenylacetyl chloride

  • MDL number MFCD08458136
  • Cas Number : 74426-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(2-Chloro-2-oxoethyl)benzotrifluoride, 4-[(Chlorocarbonyl)methyl]benzotrifluoride
Purity 95
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 94-97/10mm
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethylphenylacetyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethylphenylacetyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart