APO455838726 Apollo Scientific Ltd

3,4-Difluorophenylacetyl chloride

  • MDL number MFCD08458138
  • Cas Number : 121639-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 91-92/6mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10668

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10668
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart