APO455838728 Apollo Scientific Ltd

1-(4-Fluorobenzoyl)-4-hydroxypyrrolidine-2-carboxylic acid

  • MDL number MFCD00202775
  • Cas Number : 497163-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-208
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3243

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3243
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart