APO455838732 Apollo Scientific Ltd

2-(Cyanomethyl)-5-(2-fluorobenzoyl)thiophene

  • MDL number MFCD00664551
  • Cas Number : 303144-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-86
Synonym 2-5-(2-Fluorobenzoyl)thien-2ylacetonitrile
Purity 97
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3249

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3249
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart