APO455838744 Apollo Scientific Ltd

2-(Trifluoromethyl)phenylhydrazine hydrochloride

  • MDL number MFCD00102619
  • Cas Number : 3107-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220
Synonym 2-Hydrazinobenzotrifluoride hydrochloride
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethylphenylhydrazine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethylphenylhydrazine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart