APO455838746 Apollo Scientific Ltd

7-Amino-6-(4-fluorophenyl)-2-(methylthio)[1,2,4]triazolo[1,5-a]pyrimidine

  • MDL number MFCD00138905
  • Cas Number : 338394-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 286-288
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3430

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=3430
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart