APO455838747 Apollo Scientific Ltd

4-(4-Fluorophenyl)-2-oxo-2,5-dihydrofuran-3-carbonitrile

  • MDL number MFCD00139244
  • Cas Number : 50691-04-8
  • Molecular Weight : 203.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-212
Purity 97
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-4-fluorophenyl-2-oxo-25-dihydrofuran-3-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-fluorophenyl-2-oxo-25-dihydrofuran-3-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart